Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRS2 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10068244G
Description
IRS2 Polyclonal antibody specifically detects IRS2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
IRS2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
insulin receptor substrate 2, IRS-2 | |
The immunogen recognized by this antibody maps to a region between residue 1288 and 1338 of human insulin receptor substrate 2 using the numbering given in entry NP_003740.2 (GeneID 8660). | |
0.1 mL | |
Cell Cycle and Replication, Lipid and Metabolism, Neuroscience, Phospho Specific, Signal Transduction | |
8660 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction