Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRS4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP312966
This item is not returnable.
View return policy
Description
IRS4 Polyclonal antibody specifically detects IRS4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| IRS4 | |
| Polyclonal | |
| PBS, 1% BSA, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 8471 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| insulin receptor substrate 4,160 kDa phosphotyrosine protein, IRS-4pp160, Phosphoprotein of 160 kDa, PY160 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human IRS4. The exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction