Learn More
Invitrogen™ IRS4 Polyclonal Antibody

Description
Centrifuge briefly prior to opening.
Specifications
Specifications
| Antigen | IRS4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | IRS4 |
| Gene Accession No. | O14654 |
| Gene Alias | 160 kDa phosphotyrosine protein; Insulin receptor substrate 4; Irs4; IRS-4; Phosphoprotein of 160 kDa; pp160; py160; RP23-415G1.3 |
| Gene Symbols | IRS4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.