Learn More
IRS4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305344AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
IRS4 encodes the insulin receptor substrate 4, a cytoplasmic protein that contains many potential tyrosine and serine/threonine phosphorylation sites. Tyrosine-phosphorylated IRS4 protein has been shown to associate with cytoplasmic signalling molecules that contain SH2 domains. The IRS4 protein is phosphorylated by the insulin receptor tyrosine kinase upon receptor stimulation.Specifications
IRS4 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O14654 | |
IRS4 | |
Between 1207 and 1257 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
IRS4 | |
IRS-4, PY160 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8471 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.