Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ IRX-3, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABS1050MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS1050MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1050MI Supplier MilliporeSigma™ Supplier No. ABS1050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Iroquois-class homeodomain protein IRX-3 (UniProt P81067; also known as Homeodomain protein IRXB1, Iroquois homeobox protein 3) is encoded by the Irx3 (also known as Irxb1) gene (Gene ID 16373) in murine species. Iroquois homeobox (Irx) genes encode a family of homeodomain transcription factors that are conserved from worms to vertebrates. Six IRX genes are located in two clusters in human (IRXA and IRXB) and mouse (IrxA and IrxB) genomes. Irx3 and Irx5 of the IrxB cluster are expressed in the developing mouse limb bud and are essential for proximal and anterior skeletal formation. Irx3 and Irx5 are required to establish early anteroposterior (AP) polarity and specify the anterior progenitor population. Individual Irx genes are also important for certain postnatal cardiac functions in the developing mouse heart, including cardiac repolarization (Irx5) and rapid ventricular conduction (Irx3). Postnatal deletion of Irx3 and Irx5 in the myocardium leads to prolongation of atrioventricular conduction, due in part to activation of expression of the Na(+) channel protein Nav1.5. However, combined postnatal loss of Irx3 and Irx5 results in a restoration of the repolarization gradient that is altered in Irx5 mutant hearts, suggesting that postnatal Irx3 activity can be repressed by Irx5 in mice.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRX-3
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody serum with 0.05% Sodium Azide.
Gene Irx3, Irxb1
Gene Accession No. P81067
Host Species Rabbit
Immunogen Recombinant murine IRX-3/IRXB1 C-terminal fragment.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001240751
Test Specificity Expected to react with both spliced isoforms of murine IRX-3/IRXB1 reported by UniProt (P81067).
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.