Learn More
Description
Specifications
Specifications
| Antigen | IRX-5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Formulation | Rabbit polyclonal antibody serum with 0.05% Sodium Azide. |
| Gene | Irx5, Irxb2 |
| Gene Accession No. | Q9JKQ4 |
| Host Species | Rabbit |
| Immunogen | His-tagged recombinant murine IRX-5/IRXB2 C-terminal fragment GST fusion protein (Chen, C.W., et al. (2005). Dev. Biol. 287(1):48-60). |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.