Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRX1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS20469R

Description
The IRXA group includes Irx1, Irx2 and Irx4; the IRXB group is comprises Irx3, Irx5 and Irx6. Irx1 and Irx2 are both widely expressed during development in the lung epithelium and also in the ventricular septum. Irx1 and Irx2 also play a role in digit formation (E11.5-E14.5). The Irx gene family members are each expressed in a distinct pattern during mouse heart development. Specifically, Irx1 and Irx2 are expressed in the ventricular septum and Irx3 is expressed in the ventricular trabeculated myocardium. In addition, Irx4 is expressed in the linear heart tube and the AV canal; .Specifications
IRX1 | |
Polyclonal | |
Unconjugated | |
IRX1 | |
Homeodomain protein IRXA1; iroquois homeobox 1; Iroquois homeobox protein 1; Iroquois related homeobox 1; Iroquois related homeobox 1 (Drosophila); Iroquois-class homeodomain protein IRX-1; IRX1; IRX-5; IRXA1 | |
Rabbit | |
Protein A | |
RUO | |
79192 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
PBS with 1% BSA and 15mM sodium azide; pH | |
P78414 | |
IRX1 | |
KLH conjugated synthetic peptide derived from human IRX1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction