Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRX2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2851090.1ML
Description
IRX2 Polyclonal specifically detects IRX2 in Human samples. It is validated for Western Blot.Specifications
IRX2 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
153572 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Homeodomain protein IRXA2, iroquois homeobox 2, Iroquois homeobox protein 2, iroquois-class homeodomain protein IRX-2, IRXA2 | |
The immunogen is a synthetic peptide directed towards the middle region of Human IRX2 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction