Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549818
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549818 Supplier Invitrogen™ Supplier No. PA549818
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total IRX3 protein.

IRX3 is a member of the Iroquois homeobox gene family (seeIRX1; MIM 606197) and plays a role in an early step of neural development. Members of this family appear to play multiple roles during pattern formation of vertebrate embryos.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRX3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IRX3
Gene Accession No. P78415, P81067
Gene Alias AI894186; homeodomain protein IRXB1; iroquois homeobox 3; iroquois homeobox protein 3; Iroquois related homeobox 3; iroquois-class homeodomain protein IRX-1; iroquois-class homeodomain protein IRX-3; IRX-1; IRX3; Irxb1
Gene Symbols IRX3
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human IRX3 (aa 271-320).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16373, 79191
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.