Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568771
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568771 Supplier Invitrogen™ Supplier No. PA568771
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 86%; Mouse: 100%; Pig: 79%; Rat: 86%.

IRX3 is a member of the Iroquois homeobox gene family (seeIRX1; MIM 606197) and plays a role in an early step of neural development. Members of this family appear to play multiple roles during pattern formation of vertebrate embryos.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRX3
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IRX3
Gene Accession No. P81067
Gene Alias AI894186; homeodomain protein IRXB1; iroquois homeobox 3; iroquois homeobox protein 3; Iroquois related homeobox 3; iroquois-class homeodomain protein IRX-1; iroquois-class homeodomain protein IRX-3; IRX-1; IRX3; Irxb1
Gene Symbols IRX3
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of mouse IRX3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16373
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.