Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IRX3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144938
Description
Immunogen sequence: GEEEDTGGEG LADDDEDEEI DLENLDGAAT EPELSLAGAA RRDGDLGLGP I Antigen sequence identity: Mouse 84%; Rat 82%.
IRX3 is a member of the Iroquois homeobox gene family (seeIRX1; MIM 606197) and plays a role in an early step of neural development. Members of this family appear to play multiple roles during pattern formation of vertebrate embryos.
Specifications
IRX3 | |
Polyclonal | |
Unconjugated | |
IRX3 | |
AI894186; homeodomain protein IRXB1; iroquois homeobox 3; iroquois homeobox protein 3; Iroquois related homeobox 3; iroquois-class homeodomain protein IRX-1; iroquois-class homeodomain protein IRX-3; IRX-1; IRX3; Irxb1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79191 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P78415 | |
IRX3 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 235-285. Recombinant protein control fragment (Product #RP-109846). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction