Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Irx5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568832
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568832 Supplier Invitrogen™ Supplier No. PA568832
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 92%; Guinea Pig: 86%; Horse: 100%; Human: 93%; Mouse: 100%; Pig: 93%; Rabbit: 86%; Rat: 100%.

Establishes the cardiac repolarization gradient by its repressive actions on the KCND2 potassium-channel gene. Required for retinal cone bipolar cell differentiation. May regulate contrast adaptation in the retina and control specific aspects of visual function in circuits of the mammalian retina. Could be involved in the regulation of both the cell cycle and apoptosis in prostate cancer cells. Involved in craniofacial and gonadal development. Modulates the migration of progenitor cell populations in branchial arches and gonads by repressing CXCL12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Irx5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IRX5
Gene Accession No. Q9JKQ4
Gene Alias HMMS; Homeodomain protein IRX-2A; homeodomain protein IRXB2; iroquois homeobox 5; iroquois homeobox protein 5; Iroquois related homeobox 5 (Drosophila); iroquois-class homeodomain protein IRX-5; IRX2A; IRX-2a; IRX5; IRXB2; LOW QUALITY PROTEIN: iroquois-class homeodomain protein IRX-5
Gene Symbols IRX5
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse Irx5.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54352
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.