Learn More
Invitrogen™ IRX6 Monoclonal Antibody (1A5)

Description
Peptide Sequence: PRLTMHYPCL EKPRIWSLAH TATASAVEGA PPARPRPRSP ECRMIPGQPP ASARRLSVPR DSACDESSCI PKAFGNPKFA LQGLPLNCAP CPRRSEPVVQ CQYPSGAEAG.
Specifications
Specifications
| Antigen | IRX6 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1A5 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | IRX6 |
| Gene Accession No. | P78412 |
| Gene Alias | homeodomain protein IRXB3; iroquois homeobox 6; iroquois homeobox protein 6; iroquois homeobox protein 7; Iroquois related homeobox 6; Iroquois related homeobox 6 (Drosophila); iroquois-class homeodomain protein IRX-6; IRX-3; IRX6; IRX7; Irxb3; RGD1564830 |
| Gene Symbols | IRX6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.