Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IRX6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569494
Description
This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 92%; Horse: 85%; Human: 100%; Mouse: 86%; Rabbit: 92%; Rat: 92%.
IRX6 gene ontology annotations related to this gene include DNA-binding transcription factor activity, RNA polymerase II-specific; sequence-specific DNA binding.
Specifications
| IRX6 | |
| Polyclonal | |
| Unconjugated | |
| IRX6 | |
| homeodomain protein IRXB3; iroquois homeobox 6; iroquois homeobox protein 6; iroquois homeobox protein 7; Iroquois related homeobox 6; Iroquois related homeobox 6 (Drosophila); iroquois-class homeodomain protein IRX-6; IRX-3; IRX6; IRX7; Irxb3; RGD1564830 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 307715 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| IRX6 | |
| synthetic peptide directed towards the middle region of human IRX6. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction