Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRX6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597880
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597880 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597880 Supplier Invitrogen™ Supplier No. PA597880
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IRX6 Polyclonal Antibody for Western Blot, IHC (P), ELISA

IRX6 gene ontology annotations related to this gene include DNA-binding transcription factor activity, RNA polymerase II-specific; sequence-specific DNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRX6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene IRX6
Gene Accession No. P78412
Gene Alias homeodomain protein IRXB3; iroquois homeobox 6; iroquois homeobox protein 6; iroquois homeobox protein 7; Iroquois related homeobox 6; Iroquois related homeobox 6 (Drosophila); iroquois-class homeodomain protein IRX-6; IRX-3; IRX6; IRX7; Irxb3; RGD1564830
Gene Symbols IRX6
Host Species Rabbit
Immunogen Recombinant Human Iroquois-class homeodomain protein IRX-6 protein (289-389AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307715, 79190
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.