Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRX6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553786 Supplier Invitrogen™ Supplier No. PA553786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEEAEDEEVV STAGDRLTEF RKGAQSLPGP CAAAREGRLE RRECGLAAPR FSFNDPSGSE EADFLSAETG SPRLTMHYPC LEKPRIWSLA HTATASAVEG APPARPRPRS PECRMIP Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 62%.

IRX6 gene ontology annotations related to this gene include DNA-binding transcription factor activity, RNA polymerase II-specific; sequence-specific DNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRX6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IRX6
Gene Accession No. P78412
Gene Alias homeodomain protein IRXB3; iroquois homeobox 6; iroquois homeobox protein 6; iroquois homeobox protein 7; Iroquois related homeobox 6; Iroquois related homeobox 6 (Drosophila); iroquois-class homeodomain protein IRX-6; IRX-3; IRX6; IRX7; Irxb3; RGD1564830
Gene Symbols IRX6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IRX6. Recombinant protein control fragment (Product #RP-91320).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79190
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.