Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isatoic Anhydride 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
118-48-9
C8H5NO3
163.13
MFCD00006700
TXJUTRJFNRYTHH-UHFFFAOYSA-N
isatoic anhydride,2h-3,1-benzoxazine-2,4 1h-dione,1h-benzo d 1,3 oxazine-2,4-dione,isatoic acid anhydride,4h-3,1-benzoxazine-2,4 1h-dione,isato acid anhydride,n-carboxyanthranilic anhydride,unii-r8tfa74y4u,3,1-benzoxazine-2,4 1h-dione,anthranilic acid, n-carboxy-, cyclic anhydride
8359
1H-3,1-benzoxazine-2,4-dione
C1=CC=C2C(=C1)C(=O)OC(=O)N2
118-48-9
C8H5NO3
163.13
MFCD00006700
TXJUTRJFNRYTHH-UHFFFAOYSA-N
isatoic anhydride,2h-3,1-benzoxazine-2,4 1h-dione,1h-benzo d 1,3 oxazine-2,4-dione,isatoic acid anhydride,4h-3,1-benzoxazine-2,4 1h-dione,isato acid anhydride,n-carboxyanthranilic anhydride,unii-r8tfa74y4u,3,1-benzoxazine-2,4 1h-dione,anthranilic acid, n-carboxy-, cyclic anhydride
8359
1H-3,1-benzoxazine-2,4-dione
C1=CC=C2C(=C1)C(=O)OC(=O)N2
Melting Point 243°C
Flash Point 308°C
Packaging Plastic bottle
Color Beige to Brown
Infrared Spectrum Authentic
Quantity 100g
Assay Percent Range 98%
Beilstein 27, 264
Solubility Information Solubility in water: decomposes
Formula Weight 163.13
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material Isatoic anhydride, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE

EINECSNumber : 204-255-0

RTECSNumber : DM3100000

TSCA : TSCA