Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISCA2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5107150

Catalog No. PIPA5107150


Only null left
Add to Cart

Description

Description

Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 100%; Horse: 79%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%.

ISCA2 is involved in the assembly of mitochondrial iron-sulfur proteins. Probably involved in the binding of an intermediate of Fe/S cluster assembly.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISCA2
Polyclonal
Unconjugated
ISCA2
0710001C05Rik; 5730594E03Rik; c14_5557; HBLD1; HESB like domain containing 1; HESB-like domain-containing protein 1; iron-sulfur cluster assembly 2; iron-sulfur cluster assembly 2 homolog (S. cerevisiae); iron-sulfur cluster assembly 2 homolog, mitochondrial; ISA2; Isca2; MMDS4; RGD1563216
Rabbit
Affinity Chromatography
RUO
122961
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Q86U28
ISCA2
Synthetic peptide directed towards the middle region of human ISCA2 (aa 35-85).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.