Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ISCA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523993
Description
ISCA2 Polyclonal Antibody for Western Blot, IHC (P)
ISCA2 is involved in the assembly of mitochondrial iron-sulfur proteins. Probably involved in the binding of an intermediate of Fe/S cluster assembly.
Specifications
| ISCA2 | |
| Polyclonal | |
| Unconjugated | |
| ISCA2 | |
| 0710001C05Rik; 5730594E03Rik; c14_5557; HBLD1; HESB like domain containing 1; HESB-like domain-containing protein 1; iron-sulfur cluster assembly 2; iron-sulfur cluster assembly 2 homolog (S. cerevisiae); iron-sulfur cluster assembly 2 homolog, mitochondrial; ISA2; Isca2; MMDS4; RGD1563216 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 122961, 74316 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q86U28, Q9DCB8 | |
| ISCA2 | |
| KLH conjugated synthetic peptide between 72-101 amino acids from the central region of human ISCA2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction