Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISCA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556488 Supplier Invitrogen™ Supplier No. PA556488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGRLLTASLG PQARREASSS SPEAGEGQIR LTDSCVQRLL EITEGSEFLR LQVEGGGCSG FQYKFSLDTV INPDDRVFEQ GGARVVVDSD SLAFVKGAQV DFSQELIRSS FQVL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

ISCA2 is involved in the assembly of mitochondrial iron-sulfur proteins. Probably involved in the binding of an intermediate of Fe/S cluster assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISCA2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ISCA2
Gene Accession No. Q86U28
Gene Alias 0710001C05Rik; 5730594E03Rik; c14_5557; HBLD1; HESB like domain containing 1; HESB-like domain-containing protein 1; iron-sulfur cluster assembly 2; iron-sulfur cluster assembly 2 homolog (S. cerevisiae); iron-sulfur cluster assembly 2 homolog, mitochondrial; ISA2; Isca2; MMDS4; RGD1563216
Gene Symbols ISCA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ISCA2. Recombinant protein control fragment (Product #RP-89137).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122961
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.