Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISCA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113063
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113063 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113063 Supplier Invitrogen™ Supplier No. PA5113063
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ISCA2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

ISCA2 is involved in the assembly of mitochondrial iron-sulfur proteins. Probably involved in the binding of an intermediate of Fe/S cluster assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISCA2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ISCA2
Gene Accession No. Q86U28
Gene Alias 0710001C05Rik; 5730594E03Rik; c14_5557; HBLD1; HESB like domain containing 1; HESB-like domain-containing protein 1; iron-sulfur cluster assembly 2; iron-sulfur cluster assembly 2 homolog (S. cerevisiae); iron-sulfur cluster assembly 2 homolog, mitochondrial; ISA2; Isca2; MMDS4; RGD1563216
Gene Symbols ISCA2
Host Species Rabbit
Immunogen Recombinant Human Iron-sulfur cluster assembly 2 homolog, mitochondrial protein (9-154AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122961
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.