Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ISCU Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558463
Description
Immunogen sequence: TFGCGSAIAS SSLATEWVKG KTVEEALTIK NTDIAKELCL PPVKLHCSML AEDAIKAALA DYKLKQEPK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Iron-sulfur (Fe-S) clusters are necessary for several mitochondrial enzymes and other subcellular compartment proteins. They contain sulfur and iron, and are created via several steps that include cysteine desulfurases, iron donors, chaperones, and scaffold proteins. This gene encodes the two isomeric forms, ISCU1 and ISCU2, of the Fe-S cluster scaffold protein. Mutations in this gene have been found in patients with myopathy with severe exercise intolerance and myoglobinuria.
Specifications
ISCU | |
Polyclonal | |
Unconjugated | |
ISCU | |
2310020H20Rik; AA407971; HML; hnifU; iron-sulfur cluster assembly enzyme; iron-sulfur cluster assembly enzyme ISCU, mitochondrial; iron-sulfur cluster scaffold homolog; Iscu; IscU iron-sulfur cluster scaffold homolog; IscU iron-sulfur cluster scaffold homolog (E. coli); ISCU2; ISU2; Nifu; NifU-like N-terminal domain containing; nifU-like N-terminal domain-containing protein; NifU-like protein; NIFUN; RGD1309562 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23479 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H1K1 | |
ISCU | |
Recombinant protein corresponding to Human ISCU. Recombinant protein control fragment (Product #RP-102840). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction