Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISCU Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563455
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563455 Supplier Invitrogen™ Supplier No. PA563455
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PARLYHKKVV DHYENPRNVG SLDKTSKNVG TGLVGAPACG DVMKLQIQVD EKGKIVDARF K Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Iron-sulfur (Fe-S) clusters are necessary for several mitochondrial enzymes and other subcellular compartment proteins. They contain sulfur and iron, and are created via several steps that include cysteine desulfurases, iron donors, chaperones, and scaffold proteins. This gene encodes the two isomeric forms, ISCU1 and ISCU2, of the Fe-S cluster scaffold protein. Mutations in this gene have been found in patients with myopathy with severe exercise intolerance and myoglobinuria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISCU
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ISCU
Gene Accession No. Q9H1K1
Gene Alias 2310020H20Rik; AA407971; HML; hnifU; iron-sulfur cluster assembly enzyme; iron-sulfur cluster assembly enzyme ISCU, mitochondrial; iron-sulfur cluster scaffold homolog; Iscu; IscU iron-sulfur cluster scaffold homolog; IscU iron-sulfur cluster scaffold homolog (E. coli); ISCU2; ISU2; Nifu; NifU-like N-terminal domain containing; nifU-like N-terminal domain-containing protein; NifU-like protein; NIFUN; RGD1309562
Gene Symbols ISCU
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ISCU. Recombinant protein control fragment (Product #RP-103303).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23479
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.