Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ISCU Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572154
Description
Iron-sulfur (Fe-S) clusters are necessary for several mitochondrial enzymes and other subcellular compartment proteins. They contain sulfur and iron, and are created via several steps that include cysteine desulfurases, iron donors, chaperones, and scaffold proteins. This gene encodes the two isomeric forms, ISCU1 and ISCU2, of the Fe-S cluster scaffold protein. Mutations in this gene have been found in patients with myopathy with severe exercise intolerance and myoglobinuria.
Specifications
ISCU | |
Polyclonal | |
Unconjugated | |
ISCU | |
2310020H20Rik; HML; ISU2; NIFU; NIFUN; hnifU; IscU iron-sulfur cluster scaffold homolog; NifU-like N-terminal domain containing; iron-sulfur cluster assembly enzyme ISCU; mitochondrial; nifU-like N-terminal domain-containing protein; nifU-like protein | |
Rabbit | |
Protein A | |
RUO | |
23479, 66383 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 0.09% sodium azide | |
Q9D7P6, Q9H1K1 | |
ISCU | |
KLH conjugated synthetic peptide between 138-167 amino acids from the C-terminal region of human ISCU | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction