Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ISG15/UCRP Antibody (16D8), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
| Antigen | ISG15/UCRP |
|---|---|
| Clone | 16D8 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
Description
ISG15/UCRP Monoclonal antibody specifically detects ISG15/UCRP in Human samples. It is validated for Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
| ISG15/UCRP | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| G1P2interferon-induced 17-kDa/15-kDa protein, IFI15hUCRP, interferon, alpha-inducible protein (clone IFI-15K), Interferon-induced 15 kDa protein, Interferon-induced 17 kDa protein, interferon-stimulated protein, 15 kDa, ISG15 ubiquitin-like modifier, Ubiquitin cross-reactive protein, ubiquitin-like protein ISG15, UCRPIP17 | |
| A synthesized peptide derived from Human ISG15/UCRP [UniProt P05161] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 16D8 | |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Signal Transduction, Ubiquitin Proteasome Pathway | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 9636 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title