Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ISG15/UCRP Antibody (239), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290146V
Description
ISG15/UCRP Monoclonal specifically detects ISG15/UCRP in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
ISG15/UCRP | |
Monoclonal | |
DyLight 405 | |
G1P2interferon-induced 17-kDa/15-kDa protein, IFI15hUCRP, interferon, alpha-inducible protein (clone IFI-15K), Interferon-induced 15 kDa protein, Interferon-induced 17 kDa protein, interferon-stimulated protein, 15 kDa, ISG15 ubiquitin-like modifier, Ubiquitin cross-reactive protein, ubiquitin-like protein ISG15, UCRPIP17 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ISG15/UCRP (AAH09507.1; Met1-Ser165). | |
0.1 mL | |
Cell Biology, Signal Transduction, Ubiquitin Proteasome Pathway | |
9636 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunofluorescence | |
239 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction