Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ISG15/UCRP Antibody (OTI6C8), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271489AF594
Description
ISG15/UCRP Monoclonal specifically detects ISG15/UCRP in Human samples. It is validated for Western Blot.Specifications
ISG15/UCRP | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
ISG15 | |
Recombinant protein fragment corresponding to amino acids 1-157 of human ISG15/UCRP (NP_005092) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI6C8 | |
Western Blot | |
G1P2interferon-induced 17-kDa/15-kDa protein, IFI15hUCRP, interferon, alpha-inducible protein (clone IFI-15K), Interferon-induced 15 kDa protein, Interferon-induced 17 kDa protein, interferon-stimulated protein, 15 kDa, ISG15 ubiquitin-like modifier, Ubiquitin cross-reactive protein, ubiquitin-like protein ISG15, UCRPIP17 | |
Mouse | |
Affinity Purified | |
Cell Biology, Signal Transduction, Ubiquitin Proteasome Pathway | |
9636.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction