Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ISG15/UCRP Rabbit anti-Human, Clone: 239, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | ISG15/UCRP |
---|---|
Clone | 239 |
Dilution | Western Blot 1:2000-1:10000, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence |
Applications | Western Blot, ELISA, Immunofluorescence |
Classification | Monoclonal |
Description
ISG15/UCRP Monoclonal specifically detects ISG15/UCRP in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
ISG15/UCRP | |
Western Blot 1:2000-1:10000, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
Human | |
G1P2interferon-induced 17-kDa/15-kDa protein, IFI15hUCRP, interferon, alpha-inducible protein (clone IFI-15K), Interferon-induced 15 kDa protein, Interferon-induced 17 kDa protein, interferon-stimulated protein, 15 kDa, ISG15 ubiquitin-like modifier, Ubiquitin cross-reactive protein, ubiquitin-like protein ISG15, UCRPIP17 | |
Recombinant Human ISG15/UCRP protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
239 | |
Western Blot, ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cell Biology, Signal Transduction, Ubiquitin Proteasome Pathway | |
0.2 um filtered solution in PBS | |
9636 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title