Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISG20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530073 Supplier Invitrogen™ Supplier No. PA530073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji, mouse spleen. Predicted reactivity: Mouse (83%), Rat (82%), Pig (81%), Rhesus Monkey (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Interferon-induced antiviral exoribonuclease that acts on single-stranded RNA and also has minor activity towards single-stranded DNA. Exhibits antiviral activity against RNA viruses including hepatitis C virus (HCV), hepatitis A virus (HAV) and yellow fever virus (YFV) in an exonuclease-dependent manner. May also play additional roles in the maturation of snRNAs and rRNAs, and in ribosome biogenesis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISG20
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation 1% PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene ISG20
Gene Accession No. Q96AZ6, Q9JL16
Gene Alias 1600023I01Rik; 2010107M23Rik; 20kDa; CD25; DnaQL; estrogen-regulated transcript 45 protein; HEM45; interferon stimulated exonuclease; interferon stimulated exonuclease 20; interferon stimulated exonuclease gene 20; interferon stimulated exonuclease gene 20kDa; interferon stimulated gene 20kDa; interferon-stimulated gene 20 kDa protein; interferon-stimulated protein; ISG20; Promyelocytic leukemia nuclear body-associated protein ISG20; Protein DnaQL
Gene Symbols ISG20
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ISG20. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3669, 57444
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.