Learn More
                                    Invitrogen™ ISG20 Polyclonal Antibody
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113690
Description
Immunogen sequence: TSTDRLLWRE AKLDHCRRVS LRVLSERLLH KSIQNSLLGH SSVEDARATM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Dog: 85%; Guinea Pig: 85%; Horse: 86%; Human: 100%; Pig: 100%; Rabbit: 86%; Rat: 92%.
Interferon-induced antiviral exoribonuclease that acts on single-stranded RNA and also has minor activity towards single-stranded DNA. Exhibits antiviral activity against RNA viruses including hepatitis C virus (HCV), hepatitis A virus (HAV) and yellow fever virus (YFV) in an exonuclease-dependent manner. May also play additional roles in the maturation of snRNAs and rRNAs, and in ribosome biogenesis. [UniProt]. 
            Specifications
| ISG20 | |
| Polyclonal | |
| Unconjugated | |
| ISG20 | |
| 1600023I01Rik; 2010107M23Rik; 20kDa; CD25; DnaQL; estrogen-regulated transcript 45 protein; HEM45; interferon stimulated exonuclease; interferon stimulated exonuclease 20; interferon stimulated exonuclease gene 20; interferon stimulated exonuclease gene 20kDa; interferon stimulated gene 20kDa; interferon-stimulated gene 20 kDa protein; interferon-stimulated protein; ISG20; Promyelocytic leukemia nuclear body-associated protein ISG20; Protein DnaQL | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3669 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96AZ6 | |
| ISG20 | |
| Synthetic peptide directed towards the middle region of human ISG20. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
