Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISG20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120291
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120291 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120291 Supplier Invitrogen™ Supplier No. PA5120291
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Raji, A-549, MCF7, Mouse spleen, Rat spleen. The target is usually found in the following locations: Cajal body, Cytoplasm, Nucleus, nucleolus. Immunogen sequence: MAGSREVVAM DCEMVGLGPH RESGLARCSL VNVHGAVLYD KFIRPEGEIT DYRTRVSGVT PQHMVGATPF.

Interferon-induced antiviral exoribonuclease that acts on single-stranded RNA and also has minor activity towards single-stranded DNA. Exhibits antiviral activity against RNA viruses including hepatitis C virus (HCV), hepatitis A virus (HAV) and yellow fever virus (YFV) in an exonuclease-dependent manner. May also play additional roles in the maturation of snRNAs and rRNAs, and in ribosome biogenesis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISG20
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ISG20
Gene Accession No. Q96AZ6, Q9JL16
Gene Alias 1600023I01Rik; 2010107M23Rik; 20kDa; CD25; DnaQL; estrogen-regulated transcript 45 protein; HEM45; interferon stimulated exonuclease; interferon stimulated exonuclease 20; interferon stimulated exonuclease gene 20; interferon stimulated exonuclease gene 20kDa; interferon stimulated gene 20kDa; interferon-stimulated gene 20 kDa protein; interferon-stimulated protein; ISG20; Promyelocytic leukemia nuclear body-associated protein ISG20; Protein DnaQL
Gene Symbols ISG20
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human ISG20 (NP_002192.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293052, 3669, 57444
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.