Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISG20L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567699 Supplier Invitrogen™ Supplier No. PA567699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ISG20L2 Polyclonal Antibody for Western Blot

3'-> 5'-exoribonuclease involved in ribosome biogenesis in the processing of the 12S pre-rRNA. Displays a strong specificity for a 3'-end containing a free hydroxyl group.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISG20L2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ISG20L2
Gene Accession No. Q9H9L3
Gene Alias HSD38; HSD-38; interferon stimulated exonuclease gene 20 like 2; interferon stimulated exonuclease gene 20kDa like 2; interferon stimulated exonuclease gene 20kDa-like 2; interferon-stimulated 20 kDa exonuclease-like 2; ISG20L2
Gene Symbols ISG20L2
Host Species Rabbit
Immunogen Synthesized peptide derived from human ISG20L2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81875
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.