Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ISG20L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570829
Description
This target displays homology in the following species: Human: 100%.
3'-> 5'-exoribonuclease involved in ribosome biogenesis in the processing of the 12S pre-rRNA. Displays a strong specificity for a 3'-end containing a free hydroxyl group.
Specifications
ISG20L2 | |
Polyclonal | |
Unconjugated | |
ISG20L2 | |
HSD38; HSD-38; interferon stimulated exonuclease gene 20 like 2; interferon stimulated exonuclease gene 20kDa like 2; interferon stimulated exonuclease gene 20kDa-like 2; interferon-stimulated 20 kDa exonuclease-like 2; ISG20L2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
81875 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H9L3 | |
ISG20L2 | |
synthetic peptide directed towards the N-terminal region of human ISG20L2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction