Learn More
MP Biomedicals™ Isis DNA Polymerase 100U (1U/ul)
Isis DNA Polymerase 100U (1U/ul)
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP1EPSIS100
View Documents
MP Biomedicals™
11EPSIS100
100 u N/A N/A N/A
Description
Description
  • Ideal for high-fidelity amplification
  • Use to amplify DNA with difficult to read secondary structuresOptimal for blunt-end PCR cloning
  • The most thermostable polymerase available
  • Useful to amplify GC rich sequences
  • Isis™ DNA Polymerase is the most thermostable proofreading polymerase available for high fidelity PCR.
  • Isis DNA Pol Kit contains Isis DNA polymerase, optimized incubation buffers with and without MgCl2 and a separate MgCl2.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.