Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISL1 Monoclonal Antibody (1B1)
GREENER_CHOICE

Catalog No. PIMA515516
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515516 Supplier Invitrogen™ Supplier No. MA515516
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15516 targets ISL1 in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15516 immunogen is purified recombinant fragment of human ISL1 expressed in E. Coli. MA5-15516 detects ISL1 which has a predicted molecular weight of approximately 39kDa.

This gene encodes a member of the LIM/homeodomain family of transcription factors. The encoded protein binds to the enhancer region of the insulin gene, among others, and may play an important role in regulating insulin gene expression. The encoded protein is central to the development of pancreatic cell lineages and may also be required for motor neuron generation. Mutations in this gene have been associated with maturity-onset diabetes of the young.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISL1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1B1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene ISL1
Gene Accession No. P61371, P61372
Gene Alias Insulin gene enhancer protein ISL-1; insulin related protein; ISL LIM homeobox 1; Isl1; Isl-1; isl-1 homeobox; ISL1 transcription factor LIM/homeodomain (islet-1); ISL1 transcription factor, LIM/homeodomain; ISL1 transcription factor, LIM/homeodomain 1; isl-1=homeobox; Islet 1; ISLET1; islet-1; Lim-homeodomain protein Islet1
Gene Symbols ISL1
Host Species Mouse
Immunogen Purified recombinant fragment of human ISL1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16392, 3670
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.