Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISL1 Monoclonal Antibody (CL11651)
GREENER_CHOICE

Catalog No. PIMA544168
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544168 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544168 Supplier Invitrogen™ Supplier No. MA544168
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The antibody solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SRQLIPGDEF ALREDGLF Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a member of the LIM/homeodomain family of transcription factors. The encoded protein binds to the enhancer region of the insulin gene, among others, and may play an important role in regulating insulin gene expression. The encoded protein is central to the development of pancreatic cell lineages and may also be required for motor neuron generation. Mutations in this gene have been associated with maturity-onset diabetes of the young.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISL1
Applications Immunocytochemistry
Classification Monoclonal
Clone CL11651
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ISL1
Gene Accession No. P61371
Gene Alias Insulin gene enhancer protein ISL-1; insulin related protein; ISL LIM homeobox 1; Isl1; Isl-1; isl-1 homeobox; ISL1 transcription factor LIM/homeodomain (islet-1); ISL1 transcription factor, LIM/homeodomain; ISL1 transcription factor, LIM/homeodomain 1; isl-1=homeobox; Islet 1; ISLET1; islet-1; Lim-homeodomain protein Islet1
Gene Symbols ISL1
Host Species Mouse
Immunogen Recombinant protein corresponding to Human ISL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3670
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.