Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121000
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121000 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121000 Supplier Invitrogen™ Supplier No. PA5121000
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, SH-SY5Y, Mouse thymus, Rat thymus. The target is usually found in the following locations: Nucleus. Immunogen sequence: MGDMGDPPKK KRLISLCVGC GNQIHDQYIL RVSPDLEWHA ACLKCAECNQ YLDESCTCFV RDGKTYCKRD YIRLYGIKCA KCSIGFSKND FVMRARSKVY HIECFRCVAC SRQLIPGDEF ALREDGLFCR ADHDVVERAS LGAGDPLSPL HPARPLQMAA EPISARQPAL RPHVHKQPEK TTRVRTVLNE KQLHTLRTCY AANPRPDALM KEQLVEMTGL SPRVIRVWFQ NKRCKDKKRS IMMKQLQQQQ PNDKTNIQGM TGTPMVAASP ERHDGGLQAN PVEVQSYQPP WKVLSDFALQ SDIDQPAFQQ LVNFSEGGPG SNSTGSEVAS MSSQLPDTPN SMVASPIEA.

This gene encodes a member of the LIM/homeodomain family of transcription factors. The encoded protein binds to the enhancer region of the insulin gene, among others, and may play an important role in regulating insulin gene expression. The encoded protein is central to the development of pancreatic cell lineages and may also be required for motor neuron generation. Mutations in this gene have been associated with maturity-onset diabetes of the young.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ISL1
Gene Accession No. P61371, P61372, P61374
Gene Alias Insulin gene enhancer protein ISL-1; insulin related protein; ISL LIM homeobox 1; Isl1; Isl-1; isl-1 homeobox; ISL1 transcription factor LIM/homeodomain (islet-1); ISL1 transcription factor, LIM/homeodomain; ISL1 transcription factor, LIM/homeodomain 1; isl-1=homeobox; Islet 1; ISLET1; islet-1; Lim-homeodomain protein Islet1
Gene Symbols ISL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-349 of human Islet1 (NP_0021932).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16392, 3670, 64444
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.