Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567377 Supplier Invitrogen™ Supplier No. PA567377
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVDIIFHYPFL GAMGDHSKKK PGTAMCVGCG SQIH Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

ISL LIM Homeobox 2 (ISL2) is a gene located on chromosome 15q23 that encodes a transcription factor belonging to the LIM-homeodomain family. These transcription factors play critical roles in the regulation of gene expression during embryonic development, particularly in the differentiation and maintenance of neuronal and pancreatic cell lineages. ISL2 is specifically involved in the development of the nervous system, where it contributes to motor neuron differentiation and connectivity, as well as in the development of certain retinal structures. The protein functions by binding to specific DNA sequences, thereby influencing the transcription of target genes essential for cellular differentiation and identity. Although less studied than its paralog ISL1, ISL2's expression and roles in developmental pathways are crucial for proper organogenesis and tissue specification. Dysregulation of ISL2 has potential implications in developmental disorders and diseases involving neural and pancreatic tissues. Ongoing research aims to further elucidate its functions and potential as a therapeutic target in developmental biology and disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISL2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ISL2
Gene Accession No. Q96A47
Gene Alias 3110001N10Rik; Insulin gene enhancer protein ISL-2; insulin related protein 2 (islet 2); ISL LIM homeobox 2; ISL2; Isl-2; ISL2 transcription factor, LIM/homeodomain, (islet-2); islet 2; islet-2
Gene Symbols ISL2
Host Species Rabbit
Immunogen Recombinant Human ISL2. Recombinant protein control fragment (Product #RP-108038).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64843
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.