Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Islet-1 Antibody (4F6), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32619150UL
Description
Islet-1 Monoclonal antibody specifically detects Islet-1 in Human samples. It is validated for Western Blot, ELISASpecifications
Islet-1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
4F6 | |
Western Blot 1:500-1:5000, ELISA | |
insulin gene enhancer protein ISL-1, ISL LIM homeobox 1, Isl-1, ISL1 transcription factor, LIM/homeodomain, ISL1 transcription factor, LIM/homeodomain, (islet-1), Islet-1, ISLET1 | |
A synthesized peptide derived from Human Islet-1 [UniProt P61371] | |
50 μL | |
Diabetes Research, Lipid and Metabolism, Stem Cell Markers | |
3670 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction