Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Islet-1 Antibody (OTI2D8), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271050C
Description
Islet-1 Monoclonal specifically detects Islet-1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Islet-1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ISL1 | |
Recombinant protein fragment corresponding to amino acids 134-349 of human Islet-1 (NP_002193) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI2D8 | |
Western Blot | |
insulin gene enhancer protein ISL-1, ISL LIM homeobox 1, Isl-1, ISL1 transcription factor, LIM/homeodomain, ISL1 transcription factor, LIM/homeodomain, (islet-1), Islet-1, ISLET1 | |
Mouse | |
Affinity Purified | |
Diabetes Research, Lipid and Metabolism, Neuronal Stem Cell Markers, Stem Cell Markers | |
3670.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction