Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Islet-1 Rabbit anti-Human, Mouse, Rat, Clone: 1G9Q8, Novus Biologicals™
SDP

Catalog No. p-7246853 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165727 20 μg
NB165726 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB165727 Supplier Novus Biologicals Supplier No. NBP31531520UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Islet-1 Monoclonal antibody specifically detects Islet-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Islet-1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1G9Q8
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias insulin gene enhancer protein ISL-1, ISL LIM homeobox 1, Isl-1, ISL1 transcription factor, LIM/homeodomain, ISL1 transcription factor, LIM/homeodomain, (islet-1), Islet-1, ISLET1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human Islet-1 (P61371).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3670
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.