Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Islet-2 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298913V
Description
Islet-2 Polyclonal specifically detects Islet-2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Islet-2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
64843 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
DyLight 405 | |
FLJ10160, insulin gene enhancer protein ISL-2, ISL LIM homeobox 2, ISL2 transcription factor, LIM/homeodomain, (islet-2), islet-2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Islet-2. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction