Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISLR Monoclonal Antibody (C3), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543684
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543684 100 μL
PIMA543685 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543684 Supplier Invitrogen™ Supplier No. MA543684
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ISLR is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISLR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ISLR
Gene Accession No. O14498
Gene Alias HsT17563; immunoglobulin superfamily containing leucine rich repeat; immunoglobulin superfamily containing leucine-rich repeat protein; ISLR; Meflin; mesenchymal stromal cell- and fibroblast-expressing Linx paralogue; UNQ189/PRO215
Gene Symbols ISLR
Host Species Mouse
Immunogen Cys19-Phe428 (Accession O14498), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3671
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.