Learn More
Invitrogen™ ISLR Polyclonal Antibody

Description
Immunogen sequence: LLIPDFGKLE EGTYSCLATN ELGSAESSVD VALATPGEGG EDTLGRRFHG KAVEGKGCYT VDNEVQPSGP EDNVVIIYLS RAGNPEAAVA EGVPG Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 91%.
Specifications
Specifications
| Antigen | ISLR |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ISLR |
| Gene Accession No. | O14498 |
| Gene Alias | HsT17563; immunoglobulin superfamily containing leucine rich repeat; immunoglobulin superfamily containing leucine-rich repeat protein; ISLR; Meflin; mesenchymal stromal cell- and fibroblast-expressing Linx paralogue; UNQ189/PRO215 |
| Gene Symbols | ISLR |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.