Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISLR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547644
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547644 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547644 Supplier Invitrogen™ Supplier No. PA547644
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 50% cross-reactivity with recombinant human ISLR-2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ISLR2
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Islr2
Gene Accession No. Q5RKR3
Gene Alias B930052A04Rik; immunoglobulin superfamily containing leucine rich repeat 2; immunoglobulin superfamily containing leucine-rich repeat 2; immunoglobulin superfamily containing leucine-rich repeat protein 2; immunoglobulin superfamily containing leucine-rich repeat protein 2-like; Islr2; Kiaa1465; leucine-rich repeat domain and immunoglobulin domain containing axon extension protein; leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein; Linx; LOC100910729; Mbu-3; mKIAA1465; UNQ1885/PRO4329
Gene Symbols Islr2
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse ISLR-2 Cys20-Leu590.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320563
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.