Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ISLR2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988761
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8761 Supplier Bioss Supplier No. BS9025R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen ISLR2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Islr2
Gene Accession No. Q6UXK2
Gene Alias B930052A04Rik; immunoglobulin superfamily containing leucine rich repeat 2; immunoglobulin superfamily containing leucine-rich repeat 2; Immunoglobulin superfamily containing leucine-rich repeat protein 2; ISLR2; KIAA1465; leucine-rich repeat domain and immunoglobulin domain containing axon extension protein; Leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein; Linx; Mbu-3; mKIAA1465; UNQ1885/PRO4329
Gene Symbols Islr2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ISLR2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57611
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.