Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ISM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558729 Supplier Invitrogen™ Supplier No. PA558729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LREEEEARLL PRTHLQAELH QHGCWTVTEP AALTPGNATP PRTQEVTPLL LELQKLPELV HATLSTPNPD NQ Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 53%.

The protein encoded by this gene contains a type 1 thrombospondin domain, which is present in thrombospondin, a number of proteins involved in the complement pathway, as well as in extracellular matrix proteins. Two alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISM2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ISM2
Gene Accession No. Q6H9L7
Gene Alias Gm264; ISM2; isthmin 2; isthmin 2 homolog; isthmin 2 homolog (zebrafish); isthmin-2; PSEC0137; TAIL1; thrombospondin and AMOP containing isthmin-like 1; thrombospondin and AMOP domain-containing isthmin-like protein 1; Thrombospondin type-1 domain-containing protein 3; thrombospondin, type I domain-containing 3; thrombospondin, type I, domain containing 3; THSD3
Gene Symbols ISM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ISM2. Recombinant protein control fragment (Product #RP-98359).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145501
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.