Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

iso-Amyl Alcohol, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

123-51-3
C5H12O
88.15
PHTQWCKDNZKARW-UHFFFAOYSA-N
31260
CHEBI:15837

High quality chemicals for laboratory and specialized industrial use

Specific Gravity 1L = 0.81kg
Quantity 500mL
CAS 123-51-3
Molecular Formula C5H12O
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Packaging Narrow Mouth Amber Glass Bottle
SMILES CC(C)CCO
IUPAC Name 3-methylbutan-1-ol
Molecular Weight (g/mol) 88.15
PubChem CID 31260
ChEBI CHEBI:15837
Formula Weight 88.15
Flash Point 43.00°C
Chemical Name or Material iso-Amyl Alcohol
Grade Baker Analyzed™ ACS Reagent
Show More Show Less