Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iso-Propyl Alcohol, 25% (w/w), Reagents

Catalog No. 01334960
Change view
Click to view available options
:
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-960 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-960 Supplier Reagents Holdings Llc Supplier No. CI0009001A
Only null left
Add to Cart
Add to Cart

Iso-Propyl Alcohol, 25% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 67-63-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Isopropyl Alcohol, 2-Propanol, Propan-2-ol
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 82°C
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.9615
Percent Purity 25%
Grade Reagent
Chemical Name or Material Iso-Propyl Alcohol, 25% (w/w)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.