Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl Acetate, +99%, Pure, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
123-92-2
C7H14O2
130.19
MFCD00008946
MLFHJEHSLIIPHL-UHFFFAOYSA-N
isoamyl acetate,isopentyl acetate,isopentyl ethanoate,banana oil,pear oil,3-methylbutyl ethanoate,isoamyl ethanoate,iso-amyl acetate,3-methyl-1-butyl acetate,i-amyl acetate
31276
CHEBI:31725
3-methylbutyl acetate
CC(C)CCOC(=O)C
123-92-2
C7H14O2
130.19
MFCD00008946
MLFHJEHSLIIPHL-UHFFFAOYSA-N
isoamyl acetate,isopentyl acetate,isopentyl ethanoate,banana oil,pear oil,3-methylbutyl ethanoate,isoamyl ethanoate,iso-amyl acetate,3-methyl-1-butyl acetate,i-amyl acetate
31276
CHEBI:31725
3-methylbutyl acetate
CC(C)CCOC(=O)C
Viscosity 1.03 Pas (8.97°C)
Specific Gravity 0.874
Flash Point 25°C
Packaging Glass bottle
Quantity 250mL
Density 0.8740g/mL
Refractive Index 1.3990 to 1.4010
Melting Point -78.0°C
Boiling Point 142.0°C (756.0 mmHg)
Assay Percent Range 99+%
Linear Formula CH3CO2CH2CH2CH(CH3)2
Beilstein 02, 132
Merck Index 15, 5156
Solubility Information Solubility in water: 2mg/mL (20°C). Other solubilities: soluble in ethanol,diethyl phthalate,mineral oil,and 1,2-propanediol
Formula Weight 130.19
Percent Purity 99+%
Grade Pure
Chemical Name or Material Isoamyl acetate
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning